Key Documents
MTCP520020U
MTC Bio ProPette LE Adjustable Pipettor
capacity 2-20 μL
Synonym(s):
pipette
Select a Size
1 EA
$313.00
$313.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
$313.00
About This Item
UNSPSC Code:
41121500
NACRES:
NB.15
$313.00
Please contact Customer Service for Availability
feature
adjustable
manual
no. of channels 1
packaging
pack of 1 ea
manufacturer/tradename
MTC Bio P5200-20U
capacity
2-20 μL
Related Categories
General description
ProPette LE Single-Channel Pipettes provide maximum precision at virtually every conceivable volume. They feature a digital micrometer that is adjustable for precise volume control as well as a mechanical volume lock positioned at the top of the pipette that can be engaged or disengaged with one hand. These pipettors are lightweight and finely balanced for maximum ergonomics.
ProPette LE, 2 to 20µL
Features and Benefits
  • Adjustable with Roto-Lock
  • Light weight with reduced forces
  • Full volume range (0.1µL-10mL)
  • Superior accuracy and reproducibility
  • Outstanding user comfort
Legal Information
MTC is a trademark of MTC Bio
ProPette is a trademark of MTC Bio
Choose from one of the most recent versions:
Certificates of Analysis (COA)