Documents
MTCP4300FT
MTC Bio Filtered Pipette Tips
capacity 300 μL, sterile, pkg of 960 ea (10 x racks 96ea)
Synonym(s):
conical tube, filter tip, pipetting tip
About This Item
UNSPSC Code:
41121601
NACRES:
NB.02
material
clear
polypropylene
sterility
sterile
feature
DNase free
RNase free
packaging
pkg of 960 ea (10 x racks 96ea)
manufacturer/tradename
MTC Bio P4300-FT
capacity
300 μL
General description
Pipette Tips, Filtered, 300µL capacity, sterile, for 200µL and 300µL pipettes, 10 racks of 96 tips
Features and Benefits
  • For 300µL pipettes
  • Also fits most 200µL pipettes
  • Filtered or non-filtered
  • Pre-sterilized
  • Autoclavable box-rack
Legal Information
MTC is a trademark of MTC Bio
ProPette is a trademark of MTC Bio