MTCP1001A
MTC™ Bio Pure.Amp™ Aluminum Sealing Film
to fit standard PCR plates
Synonym(s):
PCR plate seal, aluminimum membrane, flow cytometry tube, sealing membrane
Select a Size
$119.00
Select a Size
About This Item
$119.00
Recommended Products
packaging
pack of 100 ea
manufacturer/tradename
MTC™ Bio P1001-A
Related Categories
General description
PureAmp Sealing Membranes are compatible with all standard PCR plates. Pierceable aluminum membrane guaranteed to keep evaporation rates below 5%.
Features and Benefits
- RNase, DNase, DNA, and PCR inhibitor free
- Evaporation rates ? 5%
- Inner packs of 10 membranes
- -80° to 120°C
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)