Key Documents
MTCM4030
MTC Bio Rigid Inoculating Needles
pack of 1000 ea (50 x bags 20 ea), sterile
Synonym(s):
culture loop
Select a Size
1000 EA
$109.00
$109.00
Please contact Customer Service for Availability
Select a Size
Change View
1000 EA
$109.00
About This Item
UNSPSC Code:
41161502
NACRES:
NB.15
$109.00
Please contact Customer Service for Availability
sterility
sterile
packaging
pack of 1000 ea (50 x bags 20 ea)
manufacturer/tradename
MTC Bio M4030
General description
Inoculating needles, rigid ABS, 50 sterile peel bags of 20 needles, 1000/pk
Features and Benefits
  • Sterile and disposable, no flaming
  • Uniform straking and inoculating
  • Hexagonal grip - improves control
  • Smooth loop surface - no gouging
Legal Information
MTC is a trademark of MTC Bio
Choose from one of the most recent versions:
Certificates of Analysis (COA)