MTCC2750
MTC™ Bio SpectraTube™ Centrifuge Tubes
capacity 50 mL, assorted colors, pack of 500 ea (20 x racks 25 ea, 5 colours per rack)
Synonym(s):
centrifuge tubes, conical tube
Select a Size
$309.00
Select a Size
About This Item
$309.00
Recommended Products
material
assorted colors
clear
conical bottom
polypropylene
screw cap
sterility
sterile
feature
graduation
packaging
pack of 500 ea (20 x racks 25 ea, 5 colours per rack)
manufacturer/tradename
MTC™ Bio C2750
parameter
15,000 × g speed
capacity
50 mL
diam. × L
29 mm × 117 mm
suitability
suitable for (stem cell research)
Looking for similar products? Visit Product Comparison Guide
General description
Features and Benefits
- Color coded caps for easy sample identification
- Sterile, RNase, DNase, pyrogen and cytotoxin free
- Sterile (gamma) and individually packed
- Leak Proof
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service