Key Documents
MT30671570
Sigma-Aldrich
EasyMix Magnetic Stiirer
for use with SevenDirect meters
About This Item
UNSPSC Code:
41103806
NACRES:
NB.72
Pricing and availability is not currently available.
packaging
pkg of 1 ea
manufacturer/tradename
Mettler-Toledo 30671570
compatibility
for use with SevenDirect meters
General description
The Right Stirrer
A dedicated stirrer for the SevenDirect generates key advantages: programmable stirring and a perfect fit with the EasyPlace electrode arm.
Program Consistent Conditions
Program the stirring speed from the pH meter to ensure consistent conditions for all operators. Define preconditioning and waiting periods.
Perfect Fit Saves Space
The EasyMix magnetic stirrer is a perfect fit with the EasyPlace electrode arm. Together, they form a tidy and compact setup.
Reliably Stir Large Volumes
The larger the sample, the higher the risk of inhomogeneity. Rely on the strength of the EasyMix to stir efficiently up to 10 L of sample volume.
Choose from one of the most recent versions:
Certificates of Analysis (COA)