Skip to Content
MilliporeSigma
All Photos(1)

Documents

MSST0047

Sigma-Aldrich

SILuProt SOST, Sclerostin human

recombinant, expressed in HEK 293 cells, SIL MS Protein Standard, 13C and 15N-labeled

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23201100
NACRES:
NA.12

biological source

human

Quality Level

recombinant

expressed in HEK 293 cells

tag

8-His tagged

Assay

≥98% (SDS-PAGE)

form

lyophilized powder

potency

≥98% Heavy amino acids incorporation efficiency by MS

technique(s)

mass spectrometry (MS): suitable

suitability

suitable for mass spectrometry (standard)

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... SOST(50964)

General description

SILuProt SOST is a recombinant, stable isotope-labeled human SOST which incorporates [13C6, 15N4]-Arginine and [13C6, 15N2]-Lysine. Expressed in human 293 cells, it is designed to be used as an internal standard for bioanalysis of SOST in mass-spectrometry. SILuProt SOST is a protein of 201 amino acids (including a N-terminal polyhistidine and tag), with a calculated molecular mass of 22.8 kDa.

Biochem/physiol Actions

Sclerostin is a secreted Wnt signaling antagonist produced almost exclusively by osteocytes. It can selectively inhibit Wnt/ β-catenin, suppressing the activity of osteoblasts as well as the viability of osteoblasts and osteocytes. Lower sclerostin levels are associated with lower bone mineral content and bone. It was demonstrated that greater total limb bone mineral content was significantly associated with greater circulating levels of sclerostin. In addition, circulating sclerostin is a biomarker of osteoporosis severity in long-term, chronic paraplegia. Serum sclerostin was associated significantly, independently, and positively with bone mineral density of both cortical and cancellous bone. Sclerostin is considered to be one of the factors associated with chronic kidney disease-mineral and bone disorder in hemodialysis patients.

Sequence

HHHHHHHHGGQQGWQAFKNDATEIIPELGEYPEPPPELENNKTMNRAENGGRPPHHPFETKDVSEYSCRELHFTRYVTDGPCRSAKPVTELVCSGQCGPARLLPNAIGRGKWWRPSGPDFRCIPDRYRAQRVQLLCPGGEAPRARKVRLVASCKCKRLTRFHNQSELKDFGTEAARPQKGRKPRPRARSAKANQAELENAY

Physical form

Supplied as a lyophilized powder containing phosphate buffered saline.

Legal Information

SILu is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service