Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MSST0041

Sigma-Aldrich

SILuProt HAVCR1 Hepatitis A virus cellular receptor 1 human

recombinant, expressed in HEK 293 cells, SIL MS Protein Standard, 13C and 15N-labeled

Synonym(s):

Kidney injury molecule 1 (KIM-1) human, T-cell immunoglobulin and mucin domain-containing protein 1 (TIMD-1), T-cell immunoglobulin mucin receptor 1 (TIM-1), mucin domain-containing protein 1 (TIMD-1)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23201100
NACRES:
NA.12

biological source

human

Quality Level

recombinant

expressed in HEK 293 cells

tag

8-His tagged

Assay

≥98% (SDS-PAGE)

form

lyophilized powder

potency

≥98% Heavy amino acids incorporation efficiency by MS

mol wt

calculated mol wt 31 kDa

technique(s)

mass spectrometry (MS): suitable

suitability

suitable for mass spectrometry (standard)

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... HAVCR1(26762)

General description

HAVCR1 is a trans-membrane glycoprotein that is highly expressed in kidney proximal tubular cells with kidney injury. The KIM-1 ectodomain is cleaved, released into urine, and canbe quantified as a sensitive and specific biomarker for Acute Kidney Injury (AKI) in rodents and humans. In a recent study it was demonstrated that HAVCR1 is released into the circulation where it reports kidney injury in mice, rats, and humans with AKI and chronic kidney disease (CKD). Urinary HAVCR1 and MCP-1 levels, either alone or in combination, were significantly higher than those in AKI negative patients. Importantly, the combination of HAVCR1 and MCP-1 outperformed either biomarker alone.

Immunogen

SVKVGGEAGPSVTLPCHYSGAVTSMCWNRGSCSLFTCQNGIVWTNGTHVTYRKDTRYKLLGDLSRRDVSLTIENTAVSDSGVYCCRVEHRGWFNDMKITVSLEIVPPKVTTTPIVTTVPTVTTVRTSTTVPTTTTVPTTTVPTTMSIPTTTTVLTTMTVSTTTSVPTTTSIPTTTSVPVTTTVSTFVPPMPLPRQNHEPVATSPSSPQPAETHPTTLQGAIRREPTSSPLYSYTTDGNDTVTESSDGLWNNNQTQLFLEHSLLTANTTKGDYKDDDDKGHHHHHHHHGGQ

Biochem/physiol Actions

SILuProt HAVCR1 is a recombinant, stable isotope-labeled human HAVCR1 which incorporates [13C6, 15N4]-Arginine and [13C6, 15N2]-Lysine. Expressed in human 293 cells, it is designed to be used as an internal standard for bioanalysis of HAVCR1 in mass-spectrometry. SILu Prot HAVCR1 is a protein consisting of 290 amino acids (the extracellular domain: Ser21-Gly290, and including C-terminal polyhistidine and FLAG® tags), with a calculated molecular mass of 31 kDa.

Physical form

Supplied as a lyophilized powder containing phosphate buffered saline.

Legal Information

This product is licensed under U.S. Patent No. 7,396,688 and foreign counterparts from E. I. du Pont de Nemours and Company. The purchase of this product conveys to the buyer the nontransferable right to use the purchased amount of the product for research and development only, including services for a third party for consideration. The buyer cannot sell or otherwise transfer this product, its components or materials made using this product or its components to a third party. Information about licenses for excluded uses is available from: E. I. du Pont de Nemours and Company; Attn: Associate Director, Commercial Development; DuPont Experimental Station E268; 200 Powdermill Rd.; Wilmington, DE 19803; 1-877-881-9787 (voice), 1-302-695-1437 (fax), licensing@dupont.com.
FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany
SILu is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service