Skip to Content
MilliporeSigma
All Photos(1)

Documents

MSST0036

Sigma-Aldrich

SILuLite CCL2 C-C motif chemokine 2 human

recombinant, expressed in HEK 293 cells, MS Protein Standard

Synonym(s):

HC11, Monocyte chemoattractant protein 1, Monocyte chemotactic protein 1(MCP-1), Monocyte secretory protein JE, Monocytechemoattractantprotein1, Monocytechemotacticandactivatingfactor(MCAF), Small-inducible cytokine A2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23201100
NACRES:
NA.12

biological source

human

Quality Level

recombinant

expressed in HEK 293 cells

Assay

≥98% (SDS-PAGE)

form

lyophilized powder

potency

≥98% Heavy amino acids incorporation efficiency by MS

technique(s)

mass spectrometry (MS): suitable

suitability

suitable for mass spectrometry (internal calibrator)

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... CCL2(6347)

Related Categories

General description

CCL2, the most thoroughly characterized CC chemokine, is a monocyte-specific chemoattractant that was demonstrated to attract T lymphocytes and NK cells, but not neutrophils. Enhanced expression of CCL2 was demonstrated in a variety of pathologic conditions, associated with inflammation and mononuclear cell infiltration. Extensive experimental evidence suggests that CCL2 is highly expressed in atherosclerotic plaques. In a recent study, CCL2 was shown to have value as a biomarker for risk stratification in both the initial and the chronic phases after an acute coronary syndromes (ACS). In contrast to CRP, CCL2 levels may be an indicator of long-term cardiovascular risk rather than a marker of acute prognosis. In urine, CCL2 levels were directly associated to Systemic Lupus Erythematosus disease activity, making it a non-invasive biomarker (avoiding blood sampling or tissue biopsies).

Immunogen

QPDAINAPVTCCYNFTNRKISVQRLASYRRITSSKCPKEAVIFKTIVAKEICADPKQKWVQDSMDHLDKQTQTPKTDYKDDDDKGHHHHHHHHGGQ

Biochem/physiol Actions

SILuLite CCL2 is a recombinant human protein expressed in human 293 cells. It is a homodimer consisting of 96 amino acids (including C-terminal polyhistidine and FLAG® tags), with a calculated molecular mass of 11 kDa. SILu Lite CCL2 is an analytical standard designed to be used as starting material for preparation of calibrators and controls in LC-MS applications.

Physical form

Lyophilized from a solution of phosphate buffered saline.

Legal Information

FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany
SILu is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xue Liu et al.
Genetics and molecular biology, 44(4), e20200463-e20200463 (2021-11-23)
Pain is a significant problem worldwide that affects the quality of life of patients. Dezocine is a non-addictive analgesic drug with kappa-opioid antagonist activity and has been successfully used to alleviate of postoperative pain. In addition, dezocine has an analgesic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service