Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MED-HHPM

Sigma-Aldrich

Human Hepatocyte Plating Medium

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
Pricing and availability is not currently available.

form

liquid

shipped in

wet ice

storage temp.

2-8°C

General description

Human Hepatocyte Plating Medium is DMEM-based medium (serum added in separately) for attaching cells to cultureware. Base media shipped and stored refrigerated (~ 4 °C).

Application

See technical bulletin for protocols for use with LifeNet Health® Primary Human Hepatocytes.

Legal Information

LifeNet Health is a registered trademark of LifeNet Health

configured for

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service