Skip to Content
MilliporeSigma
All Photos(3)

Documents

MAK563

Sigma-Aldrich

MPO Colorimetric Activity Assay Kit

sufficient for 100 colorimetric tests

Synonym(s):

Myeloperoxidase Activity Assay Kit

Sign Into View Organizational & Contract Pricing


About This Item

usage

sufficient for 100 colorimetric tests

Quality Level

detection method

colorimetric

relevant disease(s)

cancer; hematological disorder; cardiovascular diseases

storage temp.

−20°C

Related Categories

General description

Myeloperoxidase (MPO) is a heme-containing enzyme that catalyzes the hydrogen peroxidase-mediated oxidation of halide ions to hypochlorous acid. MPO is a lysosomal protein, highly expressed in neutrophils, that plays a role in the antimicrobial actions that occur as a result of neutrophil stimulation. The actions of MPO may also contribute to the initiation and pathogenesis of cardiovascular disease.

Application

The MPO Colorimetric Activity Assay Kit can been used to determine the activity of myeloperoxidase in cell, tissue, serum and white blood cell samples.

Features and Benefits

Click here to download a calculator excel file.
Highly sensitive, simple and high-throughput assay for measuring Myeloperoxidase (MPO) concentration.
Supportive calculator will calculate the reagents needed according to your experimental needs, and analyze your results based on your experimental data.

Suitability

Suitable for the detection of Myeloperoxidase (MPO) in various biological samples such as cells, tissue, serum and white blood cells.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

188.6 °F

Flash Point(C)

87 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service