Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAK555

Sigma-Aldrich

Lysyl Oxidase (LOX) Assay Kit

sufficient for 500 fluorometric tests

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116104
NACRES:
NA.84
Pricing and availability is not currently available.

detection method

fluorometric

relevant disease(s)

dermatological diseases; orthopedic diseases; pulmonary disorders

storage temp.

−20°C

General description

Lysyl oxidase (LOX) is an extracellular enzyme that catalyzes formation of aldehydes from lysine residues in collagen and elastin precursors. These aldehydes are highly reactive and undergo spontaneous chemical reactions with other lysyl oxidasederived aldehyde residues, or with unmodified lysine residues. This results in cross-linking collagen and elastin which is essential for stabilization of collagen fibrils and for the integrity and elasticity of mature elastin. Lysyl oxidase activity in biological samples is traditionally and most reliably assessed by tritium release end-point assays using radiolabeled collagen or elastin substrates involving laborious vacuum distillation of the released tritiated water. ​

The Lysyl Oxidase Assay Kit offers a sensitive fluorescent assay to measure LOX activity using a LOX substrate that releases hydrogen peroxide upon oxidation. The amount of hydrogen peroxide released by the LOX oxidation is detected using HRP substrate in the HRP-coupled reactions. This allows the detection of sub ng/mL lysyl oxidase. This method eliminates the interference that occurs in some biological samples and can be readily used to detect lysyl oxidase activity in cell culture samples.

Other Notes

Please note that the kit does not include the lysyl oxidase enzyme.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service