Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAK546

Sigma-Aldrich

Glycerol 3-Phosphate (G3P) Assay Kit

sufficient for 200 colorimetric tests

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116104
NACRES:
NA.84
Pricing and availability is not currently available.

detection method

colorimetric

relevant disease(s)

endocrinological disorders, diabetes; obesity; neurological disorders; cardiovascular diseases; gastrointestinal diseases

shipped in

wet ice

storage temp.

−20°C

General description

Glycerol 3-Phosphate (G3P) is an important intermediate in glycolysis metabolic pathway. Animals, fungi, and plants use G3P to produce ATP. It is used to regenerate NAD+ in brain and skeletal muscle cells. G3P has been linked to lipid imbalance diseases such as obesity. ​

The G3P Assay Kit uses a red substrate to quantify the concentration of G3P, which is proportional to the concentration of hydrogen peroxide formed in the enzyme coupling reaction cycle. The kit is an optimized "mix and read" format that is compatible with HTS applications. The limit of detection for this assay is 12.5 µM G3P in 100 µL assay volume.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service