Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAK504

Sigma-Aldrich

Coenzyme A Assay Kit

sufficient for 100 colorimetric or fluorometric tests

Synonym(s):

Assay Kit for Coenzyme A Detection

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
NACRES:
NA.84

Quality Level

detection method

colorimetric
fluorometric

relevant disease(s)

endocrinological disorders, diabetes; cancer

shipped in

wet ice

storage temp.

−20°C

General description

Coenzyme A (CoA) is involved in many important biological activities including the synthesis and oxidation of fatty acids, pyruvate oxidation in the citric acid cycle, and many others. One of CoA′s most crucial roles is the carrying and transferring of acyl groups.

Features and Benefits

  • Linear detection range: 5 - 1000 µM for the colorimetric assay and 3 - 100 µM CoA for the fluorometric assay.
  • Compatible with high-throughput handling systems.

Suitability

The kit is suitable for Coenzyme A (CoA) determination in a variety of biological samples.

Principle

The Coenzyme A Assay Kit provides a simple, two-step and high-throughput assay for measuring CoA. In this assay, the first step enzymatically converts CoA to acyl-CoA and the second step oxidizes the acyl-CoA producing an enoyl-CoA and H2O2. The resulting H2O2 reacts with a specific dye to form a pink colored product. The optical density at 570 nm or fluorescence intensity at λEx = 530 nm/λEm = 585 nm is directly proportional to the CoA concentration in the sample.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Resp. Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service