Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

MAK371

Sigma-Aldrich

Phosphatidylserine Assay Kit (Fluorometric)

sufficient for 100 fluorometric tests

Synonym(s):

PS Assay Kit (Fluorometric)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.84

usage

sufficient for 100 fluorometric tests

detection method

fluorometric

storage temp.

−20°C

General description

Phosphatidylserine (PS) is a glycerophospholipid consisting of a phosphatidyl group attached to L-serine via a phosphodiester linkage. PS is a critical component of the cellular plasma membrane and accounts for 2-15% of plasma membrane lipid composition, depending on the cell or tissue type. The highest concentrations of PS are found in neuronal tissues, which are critical for maintaining conduction velocity in myelinated neurons, as well as for higher order cognitive skills such as learning and memory. In normal, healthy cells, PS is held in the inner membrane surface (facing the cytosol) by the lipid transporter protein flippase. However, in apoptotic cells, PS molecules ‘shuffle′ between the inner and outer plasma membrane monolayers. When PS molecules flip to the extracellular (outer) surface of the cell membrane, they act as a signal for macrophages to engulf and digest the (apoptotic) cell.

Features and Benefits

Compatible with high-throughput handling systems.

Suitability

Suitable for the measurement of phosphatidylserine concentration in lipid extracts such as soft tissue homogenates (i.e. liver, brain, etc.) and cultured cell lysates (adherent or suspension cells).

Principle

The Phosphatidylserine Assay Kit allows for quantification of PS in lipid extracts of cell and tissue lysates. The assay is based on the enzymatic cleavage of PS to yield phosphatidic acid and L-serine, which is subsequently metabolized and reacts with a probe to form a stable fluorophore at λex = 538 nm/λem = 587 nm. The assay is selective for PS (other phospholipids such as phosphatidylcholine, phosphatidylethanolamine or phosphatidic acid do not interfere). The kit is high-throughput ready and highly sensitive, detecting as little as 50 pmole/well of PS (5 μM in a 10 μL sample volume).

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3 - Resp. Sens. 1

Storage Class Code

10 - Combustible liquids

Flash Point(F)

188.6 °F

Flash Point(C)

87 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service