Skip to Content
MilliporeSigma
All Photos(5)

Documents

MAK353

Sigma-Aldrich

Methanol Assay Kit

sufficient for 100 colorimetric tests

Synonym(s):

Methyl Alcohol Detection Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503

usage

sufficient for 100 colorimetric tests

detection method

colorimetric

storage temp.

−20°C

General description

Methanol is the simplest alcohol, consisting exclusively of a methyl group and a hydroxyl moiety. It is both an important industrial molecule (solvent, fuel, building block for chemical synthesis) and a biological metabolite; many bacteria generate methanol as a result of anaerobic metabolism. In humans, ingestion of large quantities of methanol is toxic and suppresses the nervous system; this is termed ‘methanol poisoning′. As such, methanol is frequently used as an additive in industrial alcohols to prevent human consumption. The toxicity of methanol is primarily due to the biological product of its metabolism, formaldehyde, which is further metabolized into formic acid. At lower concentrations, methanol poisoning can cause loss of coordination and discomfort, and at higher concentrations will lead to kidney failure, blindness, and even death. In the gut, microbial breakdown of pectin-rich foods produces small amounts of methanol. In addition, some bacteria are capable of metabolizing methane gas, generating methanol, and so monitoring methanol concentration is also relevant for industrial purposes and renewable energy research.

Suitability

Suitable for the determination of methanol concentration in biological samples (serum, plasma and urine), cell and tissue culture supernatants, fruit juice and other non-alcoholic beverages and anaerobic microbial cultures.

Principle

The Methanol Assay Kit utilizes an enzymatic mechanism by which conversion of methanol is correlated stoichiometrically with generation of a colorimetric signal that can be quantified at 450 nm. The assay shows greater than 100-fold specificity for methanol over ethanol and can detect as little as 500 pmol methanol.

replaced by

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service