Skip to Content
MilliporeSigma
All Photos(1)

Documents

MAK332

Sigma-Aldrich

Pyruvate Assay Kit

sufficient for 100 colorimetric or fluorometric tests

Synonym(s):

Pyruvic Acid Detection Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

usage

sufficient for 100 colorimetric or fluorometric tests

input

biological sample(s)

application(s)

pharmaceutical

detection method

colorimetric
fluorometric

relevant disease(s)

gastrointestinal diseases; cancer

storage temp.

−20°C

Related Categories

General description

Pyruvate is a key intermediate in cellular metabolic pathways. Pyruvate can be converted to carbohydrates via gluconeogenesis, to fatty acids or energy through acetyl-CoA, to the amino acid alanine and to ethanol. Abnormal levels of pyruvate have been linked to liver diseases and metabolic disorders. Simple, direct and automation-ready procedures for measuring pyruvate concentrations find wide applications in research and drug discovery.

Application

The Pyruvate Assay Kit may be used for:
  • Drug Discovery
  • Food and Beverage Testing
  • Neurological Disease Research

Features and Benefits

Broad Detection Range: Detect Pyruvate levels accurately across a wide range, from 2 µM to 500 µM, using only 10 μL of sample in a 96-well plate setup, ensuring comprehensive analysis with minimal sample requirement.



Simplified Process: Experience a streamlined process with the addition of only a single working reagent and a 30 minute room temperature reaction, reducing complexity and saving valuable time and effort.



Compatibility with High-Throughput Systems: Easily incorporate our kit into high-throughput handling systems, ensuring smooth and accurate processing, enhancing efficiency in your laboratory workflow.

Suitability

Suitable for pyruvate detection in biological samples and for studying the effects of drugs on pyruvate metabolism.

Principle

The Pyruvate Assay Kit uses a single Working Reagent that combines pyruvate oxidase and hydrogen peroxide determination in one step. The color intensity of the reaction product at 570nm or fluorescence intensity at λem/ex = 585/530nm is directly proportional to pyruvate concentration in the sample. Linear detection range in 96-well plate: 2 to 500 μM (17 μg/dL to 4.4 mg/dL) pyruvate for colorimetric assays and 0.2 to 50 μM for fluorometric assays.

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

188.6 °F

Flash Point(C)

87 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Varun Kamat et al.
eLife, 10 (2021-11-05)
Oxygen (O2) and other dissolved gases such as the gasotransmitters H2S, CO, and NO affect cell metabolism and function. To evaluate effects of dissolved gases on processes in tissue, we developed a fluidics system that controls dissolved gases while simultaneously

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service