Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAK299

Sigma-Aldrich

NNMT Inhibitor Screening Assay Kit

sufficient for 100 fluorometric tests

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.84

detection method

fluorometric

relevant disease(s)

endocrinological disorders, diabetes; obesity; cancer; neurological disorders

storage temp.

−70°C

General description

N′-Nicotinamide Methyltransferase (NNMT) (E.C. 2.1.1.1) catalyzes the N-methylation of nicotinamide, pyridines, and other analogues using S-adenosyl methionine (SAM) as the donor resulting in the production of 1-methylnicotinamide (MNA). NNMT plays a significant role in the regulation of metabolic pathways and is expressed at markedly high levels in several kinds of cancers, neurodegenerative diseases, obesity and diabetes, indicating it is a potential molecular target for therapy.

Suitability

Suitable for screening/studying/characterizing potential inhibitors of N′-Nicotinamide Methyltransferase (NNMT).

Principle

This NNMT inhibitor screening kit utilizes SAM as the methyl group donor and nicotinamide as the substrate. NNMT methylates nicotinamide generating S-adenosylhomocysteine (SAH) and 1-methylnicotinamide. The SAH is hydrolyzed by SAH hydrolase to form homocysteine, the free thiol group of which is detected using the Thiol Detecting Probe, generating an enhanced fluorescence signal (λex = 392 nm/ λem = 482 nm). In the presence of an NNMT inhibitor, the enzymatic activity is inhibited resulting in decreased fluorescence. This assay kit is a simple, sensitive, and rapid tool to screen potential inhibitors of NNMT.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 3 - Repr. 2 - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

Flash Point(F)

84.2 °F

Flash Point(C)

29 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service