Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAK279

Sigma-Aldrich

D-Gluconate (D-Gluconic Acid) Assay Kit (Colorimetric)

sufficient for 100 colorimetric tests

Synonym(s):

D-Gluconate (D-Gluconic Acid) Detection Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.84

detection method

colorimetric

storage temp.

−20°C

General description

D-Gluconic acid (C6H12O7) is a mild organic acid that occurs from the oxidation of glucose. It is naturally present in fruit, honey, and wine When used as a food additive, it helps regulate acidity. It is a strong chelating agent, chelating the anions of calcium, iron, aluminum, copper, and other heavy metals. D-Gluconate (C6H11O7) is the salt or ester of Gluconic Acid. Due to its low toxicity, it is widely used in pharmaceutical, food, and other industries.

Suitability

Suitable for the measurement of D-Gluconate (D-Gluconic Acid) in various samples such as tissues, plants, fruits and wine

Principle

The D-Gluconate (gluconic acid) Assay Kit is a simple and sensitive method for quantifying D-gluconate. In this assay, gluconate is utilized by glucokinase to form D-gluconate-6-phosphate and ADP producing a coupled enzyme reaction that generates a colorimetric signal (450 nm), proportional to the amount of D-gluconate present. The assay is sensitive to less than 2 μM of D-gluconate in a variety of samples.

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pinar Karagoz et al.
Biotechnology for biofuels, 13, 100-100 (2020-06-10)
Platform chemicals are essential to industrial processes. Used as starting materials for the manufacture of diverse products, their cheap availability and efficient sourcing are an industrial requirement. Increasing concerns about the depletion of natural resources and growing environmental consciousness have

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service