Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAK274

Sigma-Aldrich

Triose Phosphate Isomerase Activity Colorimetric Assay Kit

sufficient for 100 colorimetric tests

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.84

detection method

colorimetric

relevant disease(s)

neurological disorders; hematological disorder

storage temp.

−20°C

General description

Triose Phosphate Isomerase (TPI or TIM: EC 5.3.1.1) is an important enzyme for glycolysis. It reversibly interconverts dihydroxyacetone phosphate and glyceraldehyde-3-phosphate, thus maintaining the equilibrium of these two triose phosphates. TPI connects glycolysis to pentose phosphate pathway and lipid metabolism. It is a stable homodimer found in almost all organisms.

Features and Benefits

Compatible with high-throughput handling systems.

Suitability

Suitable for detection of Triose Phosphate Isomerase in animal tissues (muscle, liver, heart, kidney, etc.), cell culture (adherent or suspension cells) and human serum or plasma.

Principle

This Triose Phosphate Isomerase Activity Assay kit provides a quick and easy way for determining triose phosphate isomerase activity in a variety of samples. In this kit, triose phosphate isomerase converts dihydroxyacetone phosphate into glyceraldehyde 3-phosphate, which reacts with the enzyme mix and developer to form a colored product with strong absorbance at 450 nm. The assay is simple, sensitive, and high-throughput, and can detect triose phosphate isomerase activity as low as 40 mU/mL.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service