Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAK257

Sigma-Aldrich

Tyrosinase Inhibitor Screening Kit (Colorimetric)

sufficient for 100 colorimetric tests

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.25

application(s)

cosmetics
food and beverages

detection method

colorimetric

relevant disease(s)

dermatological diseases

storage temp.

−20°C

General description

Tyrosinase or polyphenol oxidase (EC 1.14.18.1), is an oxidoreductase that participates in the biosynthesis of melanin, a ubiquitous biological pigment found in hair, eyes, skin, etc. Inhibition of tyrosinase has been a long-time target in the skin health research, cosmetics and agricultural industries because of its role in browning reactions in skin pigmentation and during fruit harvesting and handling. Skin whitening and bleaching products utilize natural or synthetic tyrosinase inhibitors in order to lighten the skin color. Polyphenols, benzaldehyde derivatives, long-chain lipids, steroids, and natural compounds have been used as tyrosinase inhibitors.
Deficiency in tyrosinase causes albino, a condition in which the melanocytes are not able to produce melanin. This increases the risk of skin cancer.

Features and Benefits

Compatible with high-throughput handling systems.

Suitability

Suitable for screening/studying/characterizing tyrosinase inhibitors.

Principle

Tyrosinase catalyzes the oxidation of tyrosine, producing a chromophore that can be detected at 510 nm. In the presence of kojic acid, a reversible inhibitor of tyrosinase, the rate of oxidation of the substrate is decreased. The kit provides a rapid, simple, sensitive, and reliable test suitable for high-throughput screening of tyrosinase inhibitors. The assay is also adaptable to a 384 well format.

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Resp. Sens. 1

Storage Class Code

10 - Combustible liquids

Flash Point(F)

188.6 °F

Flash Point(C)

87 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

OP-25 Practical Applications of Systems Biology Bioinformatics in Translational Science and Drug Discovery.
Erhan S.
3rd INTERNATIONAL BAU-DRUG DESIGN CONGRESS, 94-94 (2015)
Vinay Bhardwaj et al.
Frontiers in medicine, 8, 617068-617068 (2021-03-09)
Chemical peeling is usually performed by dermatologists, plastic surgeons, and aestheticians for the treatment of photo-aged skin, dyspigmented skin, skin prone to acne eruption, and pre-cancerous skin lesions, etc. In this research paper, we report our investigative findings to understand

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service