Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MAK202

Sigma-Aldrich

Cathepsin S Inhibitor Screening Kit

Sufficient for 100 Fluorometric tests

Synonym(s):

CTSS Inhibitor Assay Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

detection method

fluorometric

relevant disease(s)

endocrinological disorders, diabetes; cardiovascular diseases; cancer

storage temp.

−20°C

Gene Information

human ... CTSS(1520)
mouse ... CTSS(13040)
rat ... CTSS(50654)

General description

Cathepsin S (CTSS; EC 3.4.22.27) is a lysosomal cysteine proteinase involved in the degradation of proteins to peptides required for the presentation of MHC class II molecules.1 It is associated with pathological conditions such as atherosclerosis, coronary heart disease, cancer, and insulin resistance.

Suitability

Suitable for the screening of inhibitors of Cathepsin S.

Principle

The Cathepsin S Inhibitor Screening Kit is a simple assay for screening potential inhibitors of CTSS. CTSS activity is measured by cleaving a synthetic 7-amino-4-trifluoromethylcoumarin (AFC)-based substrate to yield AFC, a fluorescent product (λex = 400/ λem = 505 nm), proportional to the enzymatic activity present.

Kit Components Only

Product No.
Description

  • CTSS Reaction Buffer

  • Cathepsin S, human

  • CTSS Substrate, Z-VVR-AFC, 10 mM

  • CTSS Inhibitor Z-FF-FMK, 1 mM

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service