Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MAK177

Sigma-Aldrich

Ceruloplasmin Activity Colorimetric Kit

Sufficient for 100 Colorimetric tests

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.25

detection method

colorimetric

relevant disease(s)

gastrointestinal diseases; neurological disorders; cancer; rheumatological diseases

storage temp.

2-8°C

Gene Information

human ... CP(1356)
mouse ... CP(12870)
rat ... CP(24268)

General description

Ceruloplasmin is a multifunctional copper-containing ferroxidase present in serum. Ceruolpasmin is the major transporter of copper in the blood, carrying more than 95% of the copper found in plasma. It uses copper to couple the oxidation of substrate with the reduction of oxygen to water. Ceruloplasmin-mediated oxidation of ferrous (Fe2+) iron to ferric (Fe3+) iron contributes to the regulation of iron efflux in cells that have transportable iron stores. Inherited loss-of-function mutations in ceruloplasmin has been linked to aceruloplasminemia. Ceruloplasmin is known to form complexes with cationic proteins during inflammation and is also involved in preventing iron-induced oxidative damage. Increased ceruloplasmin levels have been observed in celiac and Crohn′s patients. Elevated serum ceruloplasmin has also been linked to cancer, pregnancy, rheumatoid arthritis, schizophrenia, and Alzheimer′s disease, whereas its low levels have been linked to Wilson′s and Menkes′ diseases. Hence, measurement of ceruloplasmin activity in biological samples can provide useful insights into several metabolic and pathological conditions.

Application

Ceruloplasmin Activity Colorimetric Kit has been used to measure serum ceruloplasmin quantity and activity.[1]

Suitability

Suitable for the detection of ceruloplasmin activity in serum.

Principle

The Ceruloplasmin Activity Colorimetric Kit provides a fast and easy-to-use procedure for measuring ceruloplasmin activity in serum. Ceruloplasmin activity is determined using substrate oxidation, which results in a colorimetric (560 nm) product proportional to the enzymatic activity present. One unit of ceruloplasmin is the amount of enzyme that will oxidize 1 μmole of substrate per minute at 25 °C.

Kit Components Only

Product No.
Description

  • Ceruloplasmin Assay Buffer

  • Ammonium Sulfate, saturated (∼4.1 M)

  • Ceruloplasmin Substrate

  • Oxidizer (100 mM)

  • Stabilizer

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nadia Abdel-Magied et al.
Biological trace element research, 194(2), 410-422 (2019-07-18)
Low-level laser therapy (LLLT) is a type of medicine that uses laser light at low levels to activate the cellular chromophores and the initiation of cellular signaling. This study aimed to evaluate the photomodulation effect of LLL against ionizing radiation
Tai-Di Chen et al.
Genetics, 206(2), 1139-1151 (2017-04-30)
Crescentic glomerulonephritis (Crgn) is a complex disorder where macrophage activity and infiltration are significant effector causes. In previous linkage studies using the uniquely susceptible Wistar Kyoto (WKY) rat strain, we have identified multiple crescentic glomerulonephritis QTL (Crgn) and positionally cloned
Varadarajan Sudhahar et al.
Arteriosclerosis, thrombosis, and vascular biology, 39(11), 2320-2337 (2019-09-27)
Copper (Cu) is essential micronutrient, and its dysregulation is implicated in aortic aneurysm (AA) development. The Cu exporter ATP7A (copper-transporting P-type ATPase/Menkes ATPase) delivers Cu via the Cu chaperone Atox1 (antioxidant 1) to secretory Cu enzymes, such as lysyl oxidase
Jereme G Spiers et al.
Antioxidants (Basel, Switzerland), 11(1) (2022-01-22)
Essential metals such as copper, iron, and zinc are cofactors in various biological processes including oxygen utilisation, cell growth, and biomolecular synthesis. The homeostasis of these essential metals is carefully controlled through a system of protein transporters involved in the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service