Skip to Content
MilliporeSigma
All Photos(1)

Documents

MAK128

Sigma-Aldrich

Indican Assay Kit

sufficient for 100 colorimetric tests

Synonym(s):

Indoxyl Sulfate Assay

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

usage

sufficient for 100 colorimetric tests

detection method

colorimetric

relevant disease(s)

gastrointestinal diseases

storage temp.

−20°C

Related Categories

General description

Indican (indoxyl sulfate) is a putrefaction product resulting from bacterial deconjugation of tryptophan in the small intestine. Indican is water soluble and excreted through urine. Increases in urine indican can indicate protein maldigestion or malabsortion, or the overgrowth of gut anaerobic bacteria. Indican levels in urine can also increase in response to a high protein diet.

Application

Indican Assay Kit has been used to measure indican concentration in urine samples.

Features and Benefits

Compatible with high-throughput handling systems. Can be adapted for use with cuvettes.

Suitability

The Indican Assay kit provides a simple and direct procedure for measuring indican levels in urine.

Principle

This assay is based on an improved Curzon and Walsh method in which the reaction of indican with p-dimethylaminobenzaldehyde results in a colorimetric product measured at 480 nm. This assay has a linear detection range between 0.2-20 mg/dL in the 96 well plate assay.

Pictograms

CorrosionExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Met. Corr. 1 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Veit Rothhammer et al.
Nature medicine, 22(6), 586-597 (2016-05-10)
Astrocytes have important roles in the central nervous system (CNS) during health and disease. Through genome-wide analyses we detected a transcriptional response to type I interferons (IFN-Is) in astrocytes during experimental CNS autoimmunity and also in CNS lesions from patients
Type I interferons and microbial metabolites of tryptophan modulate astrocyte activity and central nervous system inflammation via the aryl hydrocarbon receptor.
Rothhammer V, et al.
Nature Medicine, 22(6), 586-586 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service