Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

MAK094

Sigma-Aldrich

Protein Carbonyl Content Assay Kit

sufficient for 100 colorimetric tests

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

usage

sufficient for 100 colorimetric tests

detection method

colorimetric

relevant disease(s)

cardiovascular diseases; neurological disorders; cancer; gastrointestinal diseases

storage temp.

2-8°C

General description

Oxidative stress results when the effectiveness of antioxidant defenses is insufficient to deal with the production of reactive oxygen species (ROS). ROS can induce damage to DNA, lipids, and proteins. The oxidation of proteins results in the production of stable carbonyl groups, which can be used as a measure of oxidative injury.

Application

Protein carbonyl content assay kit has been used to determine protein carbonyl concentration.

Suitability

Suitable for quantifying carbonyls in protein samples

Principle

The Protein Carbonyl Content Assay Kit provides a simple and direct procedure for measuring carbonyl content in a variety of biological samples. Carbonyl content is determined by the derivatization of protein carbonyl groups with 2,4-dinitrophenylhydrazine (DNPH) leading to the formation of stable dinitrophenyl (DNP) hydrazone adducts, which can be detected spectrophotometrically at 375 nm, proportional to the carbonyls present.

replaced by

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Met. Corr. 1 - Muta. 2 - Skin Corr. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohammad Arif et al.
Toxicology and industrial health, 34(4), 213-218 (2018-03-10)
Lead exposure can damage cells directly by effecting DNA or indirectly by modifying proteins and enzymes. In Bangladesh, many working children are exposed to a very high level of lead during their early life due to their involvement with lead-oriented
J O Zeitz et al.
Journal of animal physiology and animal nutrition, 102(4), 999-1011 (2018-05-01)
This study hypothesized that plasma and tissue antioxidant status of broilers is positively influenced when dietary Met concentrations exceed, and negatively when they go below NRC recommendations. In addition, different Met sources are hypothesized to affect the antioxidant defence system
Effects of chilled-then-frozen storage (up to 52 weeks) on an indicator of protein oxidation and indices of protein degradation in lamb M. longissimus lumborum.
Coombs CE, et al.
Meat Science, 135, 134-141 (2018)
Melissa M Miranda-Cruz et al.
Comparative biochemistry and physiology. Toxicology & pharmacology : CBP, 213, 19-26 (2018-07-25)
Hypoxia inducible factor-1 (HIF-1) is a transcriptional factor that induces genes involved in glucose metabolism. HIF-1 is formed by a regulatory α-subunit (HIF-1α) and a constitutive β-subunit (HIF-1β). The white spot syndrome virus (WSSV) induces a shift in glucose metabolism
UV?B radiation interacts with temperature to determine animal performance.
Ghanizadeh-Kazerouni E, et al.
Functional Ecology, 30(4), 584-595 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service