Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

MAK048

Sigma-Aldrich

Lipase Activity Assay Kit III

sufficient for 100 fluorometric tests

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

usage

sufficient for 100 fluorometric tests

detection method

fluorometric

storage temp.

−20°C

General description

The lipase family of enzymes catalyzes the cleavage of the ester bonds of lipids. In mammals, this family includes many critical members including pancreatic, hepatic, endothelial, and lipoprotein lipase. Lipases, such as pancreatic lipase, are critical for the metabolism of lipids. Lipases also play key roles in processes such as cell signaling and inflammation. Measurements of lipase activity are commonly used to screen for pancreatic injury or disease, and to monitor diseases such as cystic fibrosis, celiac disease, and Crohn′s disease.

Application


  • High-Temperature Short-Time and Holder Pasteurization of Donor Milk: Impact on Milk Composition.: This study evaluates the effects of different pasteurization methods on the composition of donor milk, focusing on its nutritional and biochemical properties (Escuder-Vieco et al., 2021).

Features and Benefits

Compatible with high-throughput handling systems.

Suitability

Suitable for the detection of lipase activity in biological samples including tissue, cells and serum

Principle

The Lipase Activity Assay Kit III provides a simple and direct procedure for measuring lipase activity in a variety of samples. Lipase activity is determined using a coupled enzyme reaction, which results in the generation of methylresorufin (λex = 529/λem = 600 nm) proportional to the enzymatic activity present. One unit of lipase is the amount of enzyme that will generate 1.0 μmole of methylresorufin from the substrate per minute at 37 °C.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Raymond Rubianto Tjandrawinata et al.
Frontiers in microbiology, 13, 825251-825251 (2022-03-18)
The anti-hypercholesterolemic effect of 11 Lactobacillus isolates was investigated in vitro by measuring remaining cholesterol in growth media, growth ability in media supplemented with cholesterol, and BSH activity. Among the selected isolates, DLBSH104, DLBSH122, and DLBSK207 have demonstrated outstanding potential
Varsha Patil et al.
Frontiers in microbiology, 12, 722229-722229 (2021-09-21)
Mycobacteria such as Mycobacterium tuberculosis, the causative agent of tuberculosis that annually kills several million people worldwide, and Mycobacterium smegmatis, the non-pathogenic fast-growing mycobacteria, require oxidative phosphorylation to meet their energy requirements. We have previously shown that deletion of one
Lin Zhu et al.
Diabetes, 67(12), 2494-2506 (2018-09-15)
In clinical trials, inhibition of cholesteryl ester transfer protein (CETP) raises HDL cholesterol levels but does not robustly improve cardiovascular outcomes. Approximately two-thirds of trial participants are obese. Lower plasma CETP activity is associated with increased cardiovascular risk in human

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service