M9938
MTSEA Biotin-XX
>95% (TLC)
Synonym(s):
2-((6-((6-((Biotinoyl)amino)hexanoyl)amino)hexanoyl)amino)ethyl methanethiosulfonate, MTSEA-Biotincapcap
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C25H45N5O6S3
Molecular Weight:
607.85
MDL number:
UNSPSC Code:
12171500
Assay
>95% (TLC)
mol wt
607.87
color
white
solubility
DMF: >10 mg/mL
DMSO: >10 mg/mL
storage temp.
−20°C
SMILES string
CS(=O)(=O)SCCNC(=O)CCCCCNC(=O)CCCCCNC(=O)CCCC[C@@H]1SC[C@@H]2NC(=O)N[C@H]12
Application
Thiol-reactive reagents for labeling proteins at cysteine residues to study structure and function,
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
D A Wagner et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 21(1), 67-74 (2001-01-11)
Photo-affinity labeling and mutagenesis studies have identified several amino acids that may contribute to the ligand binding domains of ligand-gated ion channels. These types of studies, however, only generate a one-dimensional, static description of binding site structure. In this study
M Traebert et al.
The Journal of membrane biology, 180(1), 83-90 (2001-04-04)
We have combined a functional assay, surface labeling and immunocytochemical methods to compare total and surface-exposed renal type IIa Na+/Pi cotransporter protein. The wild-type type cotransporter (NaPi-IIa) and its functionally comparable cysteine mutant S460C were expressed in Xenopus oocytes. S460C
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service