Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M9818

Sigma-Aldrich

Anti-Myopodin antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
IHC (f)
WB
clone:
polyclonal
species reactivity:
rat
citations:
5
technique(s):
immunohistochemistry (frozen sections): 1-2 μg/mL using rat skeletal muscle and rat kidney
western blot: 1-2 μg/mL using cytosolic fraction of rat skeletal muscle

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 80 kDa

species reactivity

rat

technique(s)

immunohistochemistry (frozen sections): 1-2 μg/mL using rat skeletal muscle and rat kidney
western blot: 1-2 μg/mL using cytosolic fraction of rat skeletal muscle

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Myopodin (80-95 kDa), a novel actin bundling protein, is an additional member of the synaptopodin gene family. Myopodin is expressed in skeletal and cardiac muscles. Myopodin contains one PPXY motif, multiple PXXP motifs, and a nuclear export sequence (NES). In the-disc, myopodin colocalizes with α-actinin. Myopodin, like several actin-bundling proteins, has been shown to shuttle between the nucleus and cytoplasm. It is localized in nucleus in myoblasts.

Immunogen

synthetic peptide encoding amino acids 566-585 located at the mid-region of human myopodin, conjugated to KLH. This sequence is highly conserved (77% sequence identity) in mouse myopodin and is not found in human or rat synaptopodin.

Application

Anti-Myopodin has been used in:
  • immunohistochemistry
  • immunostaining
  • western blot analysis

Biochem/physiol Actions

Myopodin directly binds to actin and contains an actin-binding site in the centre of the protein. Myopodin has actin bundling activity as shown by lantraculin - a sensitive cytosolic actin bundles and nuclear actin loops in transfected cells expressing GFP-myopodin. It binds to stress fibres in a punctuated pattern into the Z-disc during myotube differentiation. Myopodin is frequently down-regulated in invasive stages of some types of cancers, including invasive bladder tumors. Frequent complete or partial deletions of the myopodin gene have been shown to occur in 80% of invasive prostate cancer cases. Expression of myopodin induces suppression of tumor growth both in vivo and in vitro. Myopodin inhibits tumor metastasis by functioning as a tumor suppressor gene.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tumor suppressor role for myopodin in bladder cancer: loss of nuclear expression of myopodin is cell-cycle dependent and predicts clinical outcome
Sanchez-Carbayo M, et al.
Oncogene, 22(34), 5298-5298 (2003)
Interaction between importin 13 and myopodin suggests a nuclear import pathway for myopodin
Liang J, et al.
Molecular and Cellular Biochemistry, 307(1-2), 93-100 (2008)
Myopodin, a synaptopodin homologue, is frequently deleted in invasive prostate cancers
Lin F, et al.
The American Journal of Pathology, 159(5), 1603-1612 (2001)
Expression of myopodin induces suppression of tumor growth and metastasis
Jing L, et al.
The American Journal of Pathology, 164(5), 1799-1806 (2004)
Differentiation-and stress-dependent nuclear cytoplasmic redistribution of myopodin, a novel actin-bundling protein
Weins A, et al.
The Journal of Cell Biology, 155(3), 393-404 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service