Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M9545

Sigma-Aldrich

7-Methoxycoumarin-4-acetyl-Arg-Pro-Lys-Pro-Val-Glu-Nva-Trp-Arg-(2,4-dinitrophenyl)Lys amide

≥97% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C78H110N22O20
CAS Number:
Molecular Weight:
1675.84
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:

Assay

≥97% (HPLC)

form

powder

solubility

0.05 M acetic acid: 1 mg/mL, clear, yellow

UniProt accession no.

storage temp.

−20°C

SMILES string

CCC[C@H](NC(=O)[C@H](CCC(O)=O)NC(=O)[C@@H](NC(=O)[C@@H]1CCCN1C(=O)[C@H](CCCCN)NC(=O)[C@@H]2CCCN2C(=O)[C@H](CCCNC(N)=N)NC(=O)CC3=CC(=O)Oc4cc(OC)ccc34)C(C)C)C(=O)N[C@@H](Cc5c[nH]c6ccccc56)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CCCCNc7ccc(cc7[N+]([O-])=O)[N+]([O-])=O)C(N)=O

InChI

1S/C78H110N22O20/c1-5-16-53(68(106)95-58(37-45-42-88-50-18-7-6-17-48(45)50)71(109)92-54(21-12-33-86-77(81)82)69(107)90-52(67(80)105)19-9-11-32-85-51-28-25-46(99(115)116)40-61(51)100(117)118)91-70(108)55(29-30-64(102)103)93-74(112)66(43(2)3)96-73(111)60-24-15-36-98(60)76(114)57(20-8-10-31-79)94-72(110)59-23-14-35-97(59)75(113)56(22-13-34-87-78(83)84)89-63(101)38-44-39-65(104)120-62-41-47(119-4)26-27-49(44)62/h6-7,17-18,25-28,39-43,52-60,66,85,88H,5,8-16,19-24,29-38,79H2,1-4H3,(H2,80,105)(H,89,101)(H,90,107)(H,91,108)(H,92,109)(H,93,112)(H,94,110)(H,95,106)(H,96,111)(H,102,103)(H4,81,82,86)(H4,83,84,87)/t52-,53-,54-,55-,56-,57-,58-,59-,60-,66-/m0/s1

InChI key

TVZDWYYXHAIMCR-BKNSPTOHSA-N

Gene Information

mouse ... Nppa(230899)

Amino Acid Sequence

MOCAc-Arg-Pro-Lys-Pro-Val-Glu-Nva-Trp-Arg-Lys-(DNP)-NH2

Application

Substrate for matrix metalloproteinases 3 and 9.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Nagase et al.
The Journal of biological chemistry, 269(33), 20952-20957 (1994-08-19)
Members of the matrix metalloproteinase (MMP) family have been implicated in disease states such as arthritis, periodontal disease, and tumor cell invasion and metastasis. Stromelysin 1 (MMP-3) has a broad substrate specificity and participates in the activation of several MMP

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service