Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M9505

Sigma-Aldrich

Met-Phe

~98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H20N2O3S
CAS Number:
Molecular Weight:
296.39
UNSPSC Code:
12352200

Assay

~98%

storage temp.

−20°C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Fiebig et al.
Molecular and cellular endocrinology, 84(1-2), R27-R30 (1992-03-01)
High-performance liquid chromatography (HPLC) is used to detect testosterone (T)-sensitive peptides in spleen cells isolated from female C57BL/10 mice immunosuppressed against Plasmodium chabaudi malaria by T treatment. Two peaks with retention times of about 25 min and 34 min, respectively

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service