Skip to Content
MilliporeSigma
All Photos(1)

Documents

M9066

Sigma-Aldrich

Microsomes from Liver, Pooled

from rat(Sprague-Dawley), male

Synonym(s):

Liver microsome preparation

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161501
NACRES:
NA.47

biological source

rat (Sprague-Dawley)

Quality Level

form

liquid

packaging

vial of ~10 mg

shipped in

dry ice

storage temp.

−70°C

Looking for similar products? Visit Product Comparison Guide

General description

Microsomes are the fraction of "submicroscopic” particles isolated from homogenates of liver and other tissues. Microsomes are rich in ribonucleic acid (RNA).

Application

Microsomes from Liver, Pooled has been used in following studies:
  • inhibition of microsomal lipid peroxidation.
  • hydroxylation of isorhynchophylline (ISOR) in rats.

Biochem/physiol Actions

Liver microsomes are subcellular particles derived from the endoplasmic reticulum of hepatic cells. These microsomes are a rich source of drug metabolizing enzymes, including cytochrome P-450. Microsome pools from various sources are useful in the study of xenobiotic metabolism and drug interactions.
Microsomes might act as cell organelles and are actively involved in protein synthesis. Carbon monoxide-binding pigment of microsomes, named P-450, is an integral element of mixed function oxidase systems involved in the oxidative demethylation and hydroxylation of drugs and steroids. Murine liver microsomes plays a crucial role in degradation of small antimicrobial β2,2-amino acid derivatives.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Metabolism of isorhynchophylline in rats detected by LC-MS.
Wang W
J. Pharm. Pharm. Sci., 13(1), 27-37 (2010)
Role of hemoprotein P-450 in fatty acid omega-hydroxylation in a soluble enzyme system from liver microsomes.
Lu AY and Coon MJ
The Journal of Biological Chemistry, 243(6), 1331-1332 (1968)
Xiaofei Zhang et al.
ACS chemical neuroscience, 8(9), 1937-1948 (2017-06-02)
Metabotropic glutamate 2 receptors (mGlu
Liver microsomes; an integrated morphological and biochemical study.
PALADE GE
The Journal of Biophysical and Biochemical Cytology, 2(2), 171-200 (1956)
Michele Gottardi et al.
Aquatic toxicology (Amsterdam, Netherlands), 201, 11-20 (2018-06-03)
Azole fungicides, designed to halt fungal growth by specific inhibition of fungal cytochrome P450 (CYP51), inhibit cytochrome P450s involved in the metabolism of xenobiotics in several non-target organisms thus raising environmental concern. The present study investigates the degree by which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service