Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M8797

Sigma-Aldrich

Methyl 4,7,8,9-tetra-O-acetyl-2-thio-N-acetyl-α-D-neuraminic acid methyl ester

≥90% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H31NO12S
CAS Number:
Molecular Weight:
521.54
MDL number:
UNSPSC Code:
12352201

Assay

≥90% (HPLC)

storage temp.

−20°C

SMILES string

COC(=O)C1(C[C@H](OC(C)=O)[C@@H](NC(C)=O)C(O1)[C@H](OC(C)=O)[C@@H](COC(C)=O)OC(C)=O)SC

Application

N-Acetylneuraminic acid derivative (glycosyl donor) suitable for chemical sialylation of carbohydrates.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hasegawa, A., et al.
Journal of Carbohydrate Chemistry, 10, 493-493 (1991)
A facile, large-scale preparation of the methyl 2-thioglycoside of N-acetylneuraminic acid, and its usefulness for the alpha-stereoselective synthesis of sialoglycosides.
A Hasegawa et al.
Carbohydrate research, 212, 277-281 (1991-06-10)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service