Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M8779

Sigma-Aldrich

4-Maleimidobutyric acid

≥98% (titration)

Synonym(s):

γ-Maleimidobutyric acid, 4-Maleimidobutanoic acid, N-(3-Carboxypropyl)maleimide, N-Maleoyl-4-aminobutyric acid, N-Maleoyl-GABA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H9NO4
CAS Number:
Molecular Weight:
183.16
Beilstein:
1455876
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98% (titration)

form

powder

color

beige

storage temp.

2-8°C

SMILES string

OC(=O)CCCN1C(=O)C=CC1=O

InChI

1S/C8H9NO4/c10-6-3-4-7(11)9(6)5-1-2-8(12)13/h3-4H,1-2,5H2,(H,12,13)

InChI key

NCPQROHLJFARLL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Modification reagent for thiol groups in proteins

replaced by

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D H Rich et al.
Journal of medicinal chemistry, 18(10), 1004-1010 (1975-10-01)
A method for the preparation of N-maleoylamino acids and esters is reported. These compounds were shown to inhibit both the oxytocin-induced smooth muscle contraction in the isolated rat uterus and the vasopressin-induced water loss from the isolated toad bladder. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service