Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M8759

Sigma-Aldrich

3-Maleimidobenzoic acid N-hydroxysuccinimide ester

Synonym(s):

MBS

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10N2O6
CAS Number:
Molecular Weight:
314.25
Beilstein:
1505254
EC Number:
MDL number:
UNSPSC Code:
47101511
PubChem Substance ID:

mp

175-177 °C (lit.)

solubility

ethyl acetate or DMF: ≤20 mg/mL (may require addition of solvent to coupling buffer to at least 5% to maintain solubility)

storage temp.

−20°C

SMILES string

O=C1CCC(N1OC(C2=CC=CC(N3C(C=CC3=O)=O)=C2)=O)=O

InChI

1S/C15H10N2O6/c18-11-4-5-12(19)16(11)10-3-1-2-9(8-10)15(22)23-17-13(20)6-7-14(17)21/h1-5,8H,6-7H2

InChI key

LLXVXPPXELIDGQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Heterobifunctional crosslinking reagent reactive toward primary amine and sulfhydryl. Typically, coupled initially to molecules containing primary amines by amide bonds buffered at pH 7.0, followed by coupling to compounds containing sulfhydryl by thioether at same pH. Useful for preparation of enzyme immunoconjugates and hapten carrier molecule conjugates.

Other Notes

The aryl maleimide is less stable than alkyl maleimide linkers.

replaced by

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

N Sattayasai et al.
Journal of interferon research, 11(1), 41-48 (1991-02-01)
Antibodies to three conserved regions of all human interferon (IFN)-alpha 1 subtypes were raised by immunizing rabbits with short synthetic peptides coupled to a carrier. These peptides correspond to amino acid residues 29-36, 31-36, 126-131, 139-151, and 142-151 of the
D A Barton et al.
Journal of microscopy, 237(1), 79-88 (2010-01-09)
Accurate preservation of microtubule and actin microfilament arrays is crucial for investigating their roles in plant cell development. Aldehyde fixatives such as paraformaldehyde or glutaraldehyde preserve cortical microtubule arrays but, unless actin microfilaments are stabilized with drugs such as m-maleimidobenzoyl
Tatsuya Maruhashi et al.
Hypertension research : official journal of the Japanese Society of Hypertension, 38(7), 485-490 (2015-03-06)
The RhoA/Rho-associated kinase (ROCK) pathway has a key physiological role in the pathogenesis of atherosclerosis. Increased ROCK activity is associated with cardiovascular diseases. Endogenous nitric oxide (NO) has an anti-atherosclerotic effect, whereas the exogenous NO-mediated cardiovascular effect still remains controversial.
H H Keah et al.
The journal of peptide research : official journal of the American Peptide Society, 57(1), 1-10 (2001-02-13)
The synthesis of peptides containing 0, 1 and 2 cysteine residues related to the human sperm tail protein, tpx-1, is described. These synthetic peptides, following conjugation to keyhole limpet hemocyanin modified with maleimidobenzoic acid N-hydroxysuccinimide ester, were used as immunogens
T Arata
Journal of biochemistry, 109(2), 335-340 (1991-02-01)
Polymerization of G-actin in the presence of salt and phalloidin was blocked by treatment of G-actin with m-maleimidobenzoic acid N-hydroxysuccinimide ester (MBS) (designated as m-actin). The actin dimer produced by chemical crosslinking of F-actin with N,N'-p-phenylenedimaleimide did not polymerize and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service