Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M8754

Sigma-Aldrich

D-Mannose 6-phosphate barium salt hydrate

≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11BaO9P · xH2O
Molecular Weight:
395.45 (anhydrous basis)
Beilstein:
5201875
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:

biological source

synthetic (inorganic)

Assay

≥98%

form

powder

technique(s)

HPLC: suitable

impurities

≤1% (enzymatic)
≤1% total fructose 6-phosphate and glucose 6-phosphate

solvent strength

≤5%

storage temp.

−20°C

SMILES string

O.[Ba++].OC1O[C@H](COP([O-])([O-])=O)[C@@H](O)[C@H](O)[C@@H]1O

InChI

1S/C6H13O9P.Ba.H2O/c7-3-2(1-14-16(11,12)13)15-6(10)5(9)4(3)8;;/h2-10H,1H2,(H2,11,12,13);;1H2/q;+2;/p-2/t2-,3-,4+,5+,6?;;/m1../s1

InChI key

RTAFQWRLJUDBCD-FDRGOYNVSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Mannose is a monosaccharide.

Application

Mannose has been used in a study to assess extraction and radicals scavenging activity of polysaccharides. [1] It has also been used in a study to investigate monocyte recruitment during atherogenesis. [2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kui Zhong et al.
International journal of biological macromolecules, 51(4), 612-617 (2012-07-04)
Extraction variables of microwave extraction mung polysaccharides (MEMPs) from mung bean hulls were optimized by Box-Behnken design. The optimal extraction parameter of MEMP was 700 W, 70 s and 17 mL/g, and highest recovery was 60.03±2.56 mg GE/g DW, which
David W Scott et al.
Arteriosclerosis, thrombosis, and vascular biology, 32(8), e51-e59 (2012-06-23)
Upregulated expression of endothelial adhesion molecules and subsequent binding to cognate monocytic receptors are established paradigms in atherosclerosis. However, these proteins are the scaffolds, with their posttranslational modification with sugars providing the actual ligands. We recently showed that tumor necrosis

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service