Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M8543

Sigma-Aldrich

Monocyte Chemotactic Protein-3 human

≥97% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture

Synonym(s):

MCP-3

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:

biological source

human

Quality Level

recombinant

expressed in E. coli

Assay

≥97% (SDS-PAGE)

form

lyophilized powder

mol wt

9000 kDa

packaging

pkg of 10 μg

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

UniProt accession no.

storage temp.

−20°C

SMILES string

CC(=O)NC(C=O)C(O)C(OC1OC(CO)C(OC2OC(COC3OC(CO)C(O)C(O)C3OC4OC(CO)C(OC5OC(CO)C(O)C(O)C5O)C(O)C4NC(C)=O)C(O)C(OC6OC(CO)C(O)C(O)C6OC7OC(CO)C(OC8OC(CO)C(O)C(O)C8O)C(O)C7NC(C)=O)C2O)C(O)C1NC(C)=O)C(O)CO

InChI

1S/C62H104N4O46/c1-15(76)63-19(5-67)32(81)48(20(80)6-68)106-55-29(64-16(2)77)38(87)51(27(13-75)102-55)109-60-47(96)52(110-62-54(44(93)36(85)24(10-72)101-62)112-57-31(66-18(4)79)40(89)50(26(12-74)104-57)108-59-46(95)42(91)34(83)22(8-70)99-59)37(86)28(105-60)14-97-61-53(43(92)35(84)23(9-71)100-61)111-56-30(65-17(3)78)39(88)49(25(11-73)103-56)107-58-45(94)41(90)33(82)21(7-69)98-58/h5,19-62,68-75,80-96H,6-14H2,1-4H3,(H,63,76)(H,64,77)(H,65,78)(H,66,79)

InChI key

JMUPMJGUKXYCMF-UHFFFAOYSA-N

Gene Information

human ... CCL7(6354)

Physical form

Lyophilized from 30% acetonitrile and 0.1% trifluoroacetic acid containing 50 μg bovine serum albumin per 1 μg of cytokine.

Analysis Note

The biological activity is measured by its ability to chemoattract mouse BaF/3 cells transfected with human CCR2A.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Opdenakker et al.
Biochemical and biophysical research communications, 191(2), 535-542 (1993-03-15)
When human MG-63 osteosarcoma cells are triggered with IL-1 beta, they produce, among various other cytokines, also monocyte chemotactic proteins (MCPs). Homogeneous MCP-1, MCP-2 and MCP-3 were found to induce production of gelatinase B and chemotaxis of monocytes. Based on
A Ben-Baruch et al.
The Journal of biological chemistry, 270(38), 22123-22128 (1995-09-22)
Monocyte chemotactic protein-3 (MCP3) is recently identified and molecularly cloned C-C chemokine that is chemotactic for and activates a great variety of inflammatory cell types. MCP3 has been reported to interact with several C-C chemokine receptors, which can be simultaneously
M Weber et al.
Journal of immunology (Baltimore, Md. : 1950), 154(8), 4166-4172 (1995-04-15)
It has been shown that CC chemokines activate basophil and eosinophil leukocytes with different selectivities and patterns of activity. The most effective are monocyte chemotactic protein-1 (MCP-1), a potent stimulus of mediator release in basophils without effects on eosinophils, RANTES

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service