Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M8530

Sigma-Aldrich

Manganese(II) chloride tetrahydrate

suitable for plant cell culture

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MnCl2 · 4H2O
CAS Number:
Molecular Weight:
197.91
EC Number:
MDL number:
UNSPSC Code:
12352207
PubChem Substance ID:

Assay

≥99% (silver nitrate titration)

technique(s)

cell culture | plant: suitable

mp

58 °C (lit.)

SMILES string

Cl[Mn]Cl.[H]O[H].[H]O[H].[H]O[H].[H]O[H]

InChI

1S/2ClH.Mn.4H2O/h2*1H;;4*1H2/q;;+2;;;;/p-2

InChI key

CNFDGXZLMLFIJV-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - STOT RE 2

Target Organs

Brain

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

does not flash

Flash Point(C)

does not flash

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lisha Luo et al.
NMR in biomedicine, 25(12), 1360-1368 (2012-05-11)
The aim of this study was to provide data on the dose dependence of manganese-enhanced MRI (MEMRI) in the visual pathway of experimental rats and to study the toxicity of MnCl₂ to the retina. Sprague-Dawley rats were intravitreally injected with
Marta Sidoryk-Wegrzynowicz et al.
Journal of neurochemistry, 122(4), 856-867 (2012-06-20)
Manganese (Mn) has been implicated in the impairment of the glutamate-glutamine cycling (GGC) by deregulation of Glu and glutamine (Gln) turnover in astrocytes. Here, we have examined possible mechanisms involved in the Mn(II)-mediated disruption of Glu turnover, including those related
Barry J Bowman et al.
Eukaryotic cell, 11(11), 1362-1370 (2012-09-18)
The pmr gene is predicted to encode a Ca(2+)-ATPase in the secretory pathway. We examined two strains of Neurospora crassa that lacked PMR: the Δpmr strain, in which pmr was completely deleted, and pmr(RIP), in which the gene was extensively
Katharina A Sterenczak et al.
BMC cancer, 12, 284-284 (2012-07-13)
Cell lines represent a key tool in cancer research allowing the generation of neoplasias which resemble initial tumours in in-vivo animal models. The characterisation of early tumour development is of major interest in order to evaluate the efficacy of therapeutic
A G Kanthasamy et al.
Toxicology letters, 214(3), 288-295 (2012-09-22)
The role of normal cellular prion protein (PrP) remains to be fully elucidated; however, the protein is crucial for the infection and progression of prion diseases. Recent evidence indicates that PrP is a metalloprotein since the octapeptide repeat sequences in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service