Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M8433

Sigma-Aldrich

Anti-phospho-c-Myc (pThr58/pSer62) antibody produced in rabbit

affinity isolated antibody, liquid

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

liquid

mol wt

antigen ~64-67 kDa

species reactivity

rat, human, mouse

technique(s)

immunohistochemistry: 10-20 μg/mL
immunoprecipitation (IP): 2-4 μg/mL
western blot: 0.5-4 μg/mL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pThr58/pSer62)

Gene Information

human ... MYC(4609)
mouse ... Myc(17869)
rat ... Myc(24577)

General description

The transcription factor c-Myc is a short-lived nuclear phosphoprotein involved in cell proliferation, differentiation, and cancer. Expression of c-Myc is strictly controlled by mitogens and is required for cell cycle entry. c-Myc binds to DNA and activates transcription as a heterodimeric complex with Max. c-Myc is phosphorylated in vitro at Ser62 by MAP Kinase, and in vivo at both Thr58 and Ser62 although the pathways and kinases acting in vivo have not been identified. Mutation of Thr58 and Ser62 to Ala inhibits the ability of c-Myc to stimulate transcription. c-Myc, a ~45 kDa protein that migrates with an apparent molecular weight of ~64-67kDa in reducing SDS-PAGE, appears to regulate multiple aspects of growth control; c-Myc promotes cell proliferation in the presence of Cdk inhibitors and induces apoptosis upon overexpression in serum deprived cells.

Specificity

Does not react with non-phosphorylated c-Myc.

Immunogen

synthetic peptide corresponding to residues around phospho-Thr58 and phospho-Ser62 of human c-Myc.

Physical form

In phosphate buffered saline (PBS), pH 7.2, containing 50% glycerol, 1% BSA, 0.02% thimerosal.

Preparation Note

Purified by protein A and peptide affinity chromatography.

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enrico Alessio et al.
Nucleic acids research, 47(4), 1653-1670 (2019-01-17)
Long non-coding RNAs (lncRNAs) are emerging as important players in the regulation of several aspects of cellular biology. For a better comprehension of their function, it is fundamental to determine their tissue or cell specificity and to identify their subcellular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service