Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M8267

Sigma-Aldrich

des-Ac-α-Melanocyte Stimulating Hormone

≥97% (HPLC)

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
US$115.00
1 KG
US$180.00

US$115.00


Availability

Estimated to ship onMarch 07, 2025



Select a Size

Change View
250 G
US$115.00
1 KG
US$180.00

About This Item

Empirical Formula (Hill Notation):
C75H107N21O18S
CAS Number:
Molecular Weight:
1622.85
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

US$115.00


Availability

Estimated to ship onMarch 07, 2025


Assay

≥97% (HPLC)

form

solid

UniProt accession no.

shipped in

ambient

storage temp.

−20°C

SMILES string

CSCCC(NC(=O)C(CO)NC(=O)C(Cc1ccc(O)cc1)NC(=O)C(N)CO)C(=O)NC(CCC(O)=O)C(=O)NC(Cc2cnc[nH]2)C(=O)NC(Cc3ccccc3)C(=O)NC(CCCNC(N)=N)C(=O)NC(Cc4c[nH]c5ccccc45)C(=O)NCC(=O)NC(CCCCN)C(=O)N6CCCC6C(=O)NC(C(C)C)C(N)=O

InChI

1S/C75H107N21O18S/c1-41(2)62(63(78)103)95-73(113)59-19-12-29-96(59)74(114)53(17-9-10-27-76)86-60(100)37-84-65(105)56(33-44-35-83-49-16-8-7-15-47(44)49)92-66(106)50(18-11-28-82-75(79)80)87-69(109)55(31-42-13-5-4-6-14-42)91-71(111)57(34-45-36-81-40-85-45)93-67(107)51(24-25-61(101)102)88-68(108)52(26-30-115-3)89-72(112)58(39-98)94-70(110)54(90-64(104)48(77)38-97)32-43-20-22-46(99)23-21-43/h4-8,13-16,20-23,35-36,40-41,48,50-59,62,83,97-99H,9-12,17-19,24-34,37-39,76-77H2,1-3H3,(H2,78,103)(H,81,85)(H,84,105)(H,86,100)(H,87,109)(H,88,108)(H,89,112)(H,90,104)(H,91,111)(H,92,106)(H,93,107)(H,94,110)(H,95,113)(H,101,102)(H4,79,80,82)

InChI key

NMMSXFAQJQPZOI-UHFFFAOYSA-N

Gene Information

Amino Acid Sequence

Ser-Tyr-Ser-Met-Glu-His-Phe-Arg-Trp-Gly-Lys-Pro-Val-NH2

Biochem/physiol Actions

Retains ~10% of the activity of α-MSH.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Need A Sample COA?

This is a sample Certificate of Analysis (COA) and may not represent a recently manufactured lot of this specific product.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service