Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M8045

Sigma-Aldrich

rac-2-Methoxy-3-octadecanamido-1-propylphosphocholine

>96% (TLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H57N2O6P
CAS Number:
Molecular Weight:
536.73
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

>96% (TLC)

impurities

≤0.5 mol/mol (NH4)2CO3

functional group

methoxy
phospholipid

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCCCC(=O)NCC(COP([O-])(=O)OCC[N+](C)(C)C)OC

InChI

1S/C27H57N2O6P/c1-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-27(30)28-24-26(33-5)25-35-36(31,32)34-23-22-29(2,3)4/h26H,6-25H2,1-5H3,(H-,28,30,31,32)

InChI key

KTJPWDKOKGANNX-UHFFFAOYSA-N

Biochem/physiol Actions

Protein kinase C inhibitor.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M H Marx et al.
Journal of medicinal chemistry, 31(4), 858-863 (1988-04-01)
Previously unreported analogues of the synthetic antitumor phospholipid ET-18-OMe (1-octadecyl-2-methoxy-rac-glycero-3-phosphocholine), in which the 1-ether oxygen has been replaced by an amido group, have been prepared and evaluated for in vitro cytotoxic effects and for inhibition of protein kinase C. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service