Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M7892

Sigma-Aldrich

[Val-OH13]-α-MSH

≥97% (HPLC)

Synonym(s):

N-Acetyl-Ser-Tyr-Ser-Met-Glu-His-Phe-Arg-Trp-Gly-Lys-Pro-Val

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C77H108N20O20S
CAS Number:
Molecular Weight:
1665.87
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥97% (HPLC)

storage temp.

−20°C

SMILES string

CSCCC(NC(=O)C(CO)NC(=O)C(Cc1ccc(O)cc1)NC(=O)C(CO)NC(C)=O)C(=O)NC(CCC(O)=O)C(=O)NC(Cc2cnc[nH]2)C(=O)NC(Cc3ccccc3)C(=O)NC(CCCNC(N)=N)C(=O)NC(Cc4c[nH]c5ccccc45)C(=O)NCC(=O)NC(CCCCN)C(=O)N6CCCC6C(=O)NC(C(C)C)C(O)=O

InChI

1S/C77H108N20O20S/c1-42(2)64(76(116)117)96-74(114)61-20-13-30-97(61)75(115)54(18-10-11-28-78)87-62(102)38-84-65(105)57(34-46-36-83-50-17-9-8-16-49(46)50)93-66(106)51(19-12-29-82-77(79)80)88-69(109)55(32-44-14-6-5-7-15-44)91-71(111)58(35-47-37-81-41-85-47)94-67(107)52(25-26-63(103)104)89-68(108)53(27-31-118-4)90-73(113)60(40-99)95-70(110)56(33-45-21-23-48(101)24-22-45)92-72(112)59(39-98)86-43(3)100/h5-9,14-17,21-24,36-37,41-42,51-61,64,83,98-99,101H,10-13,18-20,25-35,38-40,78H2,1-4H3,(H,81,85)(H,84,105)(H,86,100)(H,87,102)(H,88,109)(H,89,108)(H,90,113)(H,91,111)(H,92,112)(H,93,106)(H,94,107)(H,95,110)(H,96,114)(H,103,104)(H,116,117)(H4,79,80,82)

InChI key

MBBQINYFFIKILA-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W Barcellini et al.
Journal of leukocyte biology, 68(5), 693-699 (2000-11-10)
The purpose of the present research was to determine if alpha-melanocyte-stimulating hormone (alpha-MSH) and its C-terminal tripeptide [alpha-MSH (11-13), KPV] alter HIV expression in infected cells. The results indicate that chronically HIV-1-infected promonocytic U1 cells produce alpha-MSH and that immunoneutralization
T Watanabe et al.
Brain research bulletin, 32(3), 311-314 (1993-01-01)
Interleukin-1 (IL-1) is a proinflammatory cytokine, alpha-MSH(1-13) molecules inhibit inflammation induced by cytokines, other mediators of inflammation, and by peripheral irritants. D-valine substitution in the antiinflammatory/antipyretic message sequence [alpha-MSH(11-13), Lys-Pro-Val] of alpha-MSH(1-13) increases the activity of the tripeptide. Our aim

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service