Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M7812

Greiner 96 well 1 x 8 strip plates

high binding, flat bottom clear polystyrene wells (8 well strips)

Synonym(s):

96 multiwell plates, 96 well microplates, 96 well microtiter plates, 96 well strip plates

Sign Into View Organizational & Contract Pricing

Select a Size

100 EA
$1,200.00

$1,200.00


Please contact Customer Service for Availability


Select a Size

Change View
100 EA
$1,200.00

About This Item

UNSPSC Code:
41122107
NACRES:
NB.22
material:
clear polystyrene
flat bottom clear polystyrene wells (8 well strips)
size:
96 wells
sterility:
non-sterile
binding type:
high binding surface
feature:
lid: no

$1,200.00


Please contact Customer Service for Availability

material

clear polystyrene
flat bottom clear polystyrene wells (8 well strips)

sterility

non-sterile

feature

lid: no

packaging

case of 100

manufacturer/tradename

Greiner 762071

maximum volume

388 μL

size

96 wells

working volume

350 μL

color

clear

binding type

high binding surface

Looking for similar products? Visit Product Comparison Guide

General description

MICROLON® 600, 96W 12xF8 Strip Microplate, working volume 20-350 μL; total well volume 388 μL.

Other Notes

96 well flat (F) solid bottom microplate in white frame, F8 strips mounted in frame, high-binding, no lid, alphanumeric well coding, length: 127.76 mm; width: 85.48 mm.

Suitability

UsefμL in immunological assays. MICROLON 600 is a hydrophobic medium binding surface that will bind up to 600 ng/cm2 of non-polar proteins and peptides. Free of detectable DNase/RNase, human DNA and pyrogens.

Legal Information

MICROLON is a registered trademark of Greiner Bio-One GmbH

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service