Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M7774

Sigma-Aldrich

Magnesium sulfate heptahydrate

suitable for plant cell culture

Synonym(s):

Epsom salts

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MgSO4 · 7H2O
CAS Number:
Molecular Weight:
246.47
EC Number:
MDL number:
UNSPSC Code:
12352207
eCl@ss:
38030409
PubChem Substance ID:

vapor density

<0.01 (vs air)

vapor pressure

<0.1 mmHg ( 20 °C)

technique(s)

cell culture | plant: suitable

SMILES string

O.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O

InChI

1S/Mg.H2O4S.7H2O/c;1-5(2,3)4;;;;;;;/h;(H2,1,2,3,4);7*1H2/q+2;;;;;;;;/p-2

InChI key

WRUGWIBCXHJTDG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gabriele Fluhr et al.
BMC gastroenterology, 13, 11-11 (2013-01-17)
Acute pancreatitis is the most common complication of diagnostic and therapeutic endoscopic retrograde cholangiopancreatography (ERCP). In spite of continuing research, no pharmacologic agent capable of effectively reducing the incidence of ERCP-induced pancreatitis has found its way into clinical practise. A
Nidhi B Panda et al.
Journal of clinical anesthesia, 25(2), 92-97 (2013-01-22)
To study the minimal effective dose of magnesium sulfate to control blood pressure (BP) during intubation in hypertensive patients. Prospective, randomized, double-blind study. Operating room of an academic medical center. 80 adult, ASA physical status 1 and 2, controlled hypertensive
Sarika Chaturvedi et al.
Journal of health, population, and nutrition, 31(1), 86-95 (2013-04-27)
Severe pre-eclampsia and eclampsia are common causes of maternal deaths worldwide and more so in developing countries. Magnesium sulphate (MgSO4) is now the most-recommended drug of choice to treat these conditions. Despite favourable policies for the use of MgSO4 treatment
A P Morrison et al.
British journal of anaesthesia, 110(5), 702-712 (2013-03-28)
Spinal anaesthesia is the primary anaesthetic technique for many types of surgery. Adjuncts to the local anaesthetics (LA) used in spinal anaesthesia can exhibit undesirable side-effects, limiting their use, but magnesium may have advantages in this respect. We sought randomized
Karen J Gibbins et al.
Obstetrics and gynecology, 121(2 Pt 1), 235-240 (2013-01-25)
Clinical trials support the efficacy and safety of magnesium sulfate for cerebral palsy prevention. We evaluated the implementation of a clinical protocol for the use of magnesium for cerebral palsy prevention in our large women's hospital, focusing on uptake, indications

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service