Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M7684

Sigma-Aldrich

MRS 2159

>98% (HPLC), solid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H11N3O8PNa3
CAS Number:
Molecular Weight:
461.21
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
To inquire about this product M7684, please contact your local MilliporeSigma office or dealer. Contact Technical Service

Assay

>98% (HPLC)

form

solid

solubility

H2O: >10 mg/mL

storage temp.

2-8°C

SMILES string

[Na+].[Na+].[Na+].[H]C(=O)c1c(O)c(C)nc(\N=N\c2ccc(cc2)C([O-])=O)c1COP([O-])([O-])=O

InChI

1S/C15H14N3O8P.3Na/c1-8-13(20)11(6-19)12(7-26-27(23,24)25)14(16-8)18-17-10-4-2-9(3-5-10)15(21)22;;;/h2-6,20H,7H2,1H3,(H,21,22)(H2,23,24,25);;;/q;3*+1/p-3/b18-17+;;;

InChI key

VCUDBCPCDKEAKO-GLCFPVLVSA-K

Gene Information

human ... P2RX1(5023)

Biochem/physiol Actions

P2X1 purinoceptor antagonist.

Features and Benefits

This compound is featured on the P2 Receptors: P2X Ion Channel Family page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Legal Information

Sold under license from the National Institutes of Health

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yong-Chul Kim et al.
Drug development research, 45(2), 52-66 (1998-10-01)
Novel analogs of the P2 receptor antagonist pyridoxal-5'-phosphate-6-phenylazo-2',4'-disulfonate (PPADS) were synthesized. Modifications were made through functional group substitution on the sulfophenyl ring and at the phosphate moiety through the inclusion of phosphonates, demonstrating that a phosphate linkage is not required

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service