Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M7127

Sigma-Aldrich

1,4-Bis(2-methylstyryl)benzene

crystalline

Synonym(s):

Bis-MSB

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3C6H4CH=CH)2C6H4
CAS Number:
Molecular Weight:
310.43
Beilstein:
2375845
EC Number:
MDL number:
UNSPSC Code:
12352103

form

crystalline

color

green

mp

180-182 °C (lit.)

storage temp.

−20°C

SMILES string

Cc1ccccc1\C=C\c2ccc(cc2)\C=C\c3ccccc3C

InChI

1S/C24H22/c1-19-7-3-5-9-23(19)17-15-21-11-13-22(14-12-21)16-18-24-10-6-4-8-20(24)2/h3-18H,1-2H3/b17-15+,18-16+

InChI key

QKLPIYTUUFFRLV-YTEMWHBBSA-N

Looking for similar products? Visit Product Comparison Guide

comparable product

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 4 - Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service