Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M7034

Sigma-Aldrich

NG-Methyl-D-arginine acetate salt

≥98% (TLC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C7H16N4O2 · CH3CO2H
CAS Number:
Molecular Weight:
248.28
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98% (TLC)

SMILES string

CC(O)=O.CNC(=N)NCCC[C@@H](N)C(O)=O

Looking for similar products? Visit Product Comparison Guide

Application

No significant effect on NOS; used as a negative control in studies of L-NMMA activity.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service