Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

M7031

Sigma-Aldrich

13-Methylmyristic acid

≥98% (capillary GC)

Synonym(s):

13-Methyltetradecanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H30O2
CAS Number:
Molecular Weight:
242.40
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

≥98% (capillary GC)

form

solid

functional group

carboxylic acid

shipped in

ambient

storage temp.

2-8°C

SMILES string

CC(C)CCCCCCCCCCCC(O)=O

InChI

1S/C15H30O2/c1-14(2)12-10-8-6-4-3-5-7-9-11-13-15(16)17/h14H,3-13H2,1-2H3,(H,16,17)

InChI key

ZOCYQVNGROEVLU-UHFFFAOYSA-N

Biochem/physiol Actions

13-Methyltetradecanoic acid (13-Methylmyristic acid, 13-MTD, 13-MTDA), an iso-fatty acid, is used to study the induction of mitochondrial-mediated apoptosis via the AKT and MAPK pathways. 13-MTDA is used for the biosynthesis of methyl-branched polyhydroxyalkanoates.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tianxin Lin et al.
Urologic oncology, 30(3), 339-345 (2010-09-17)
13-Methyltetradecanoic acid (13-MTD), a saturated branched-chain fatty acid purified from soy fermentation products, is known to induce apoptosis in many types of human cancer cells. This study was designed to investigate the molecular mechanisms involved in 13-MTD-induced apoptosis in human
Michael W Ring et al.
The Journal of biological chemistry, 281(48), 36691-36700 (2006-09-23)
Iso-fatty acids (FAs) are the dominant FA family in all myxobacteria analyzed. Furthermore, it was postulated that iso-FAs or compounds derived thereof are involved in fruiting body formation in Myxococcus xanthus, since mutants with a reduced level of iso-FA due
Sawitree Wongtangtintharn et al.
Journal of nutritional science and vitaminology, 50(2), 137-143 (2004-07-10)
This paper describes the antitumoral activity of branched-chain fatty acid (BCFA) in human breast cancer cells with an emphasis on its effect on fatty acid biosynthesis. First, the relationship between chain-length and antitumoral activity was studied. The highest activity was
R A Klein et al.
Lipids, 15(8), 572-579 (1980-08-01)
We show in this paper that 13-methyltetradecanoic acid (13-MTD) can be used as a structually labeled marker for investigating the mobility of fatty acyl chains in adipose tissue in the rat. The presence of an omega-1 methyl group allows easy
S T Faung et al.
Thrombosis research, 81(1), 91-100 (1996-01-01)
Flavobacterium ranacida consisted of 75% of 13-methyl myristate in total fatty acids. The acid at > 60 microM caused the lysis of gel-filtered platelets (GFP) in both time- and concentration-dependent manners. Scanning electron microscopy showed that: 1). GFP in 40

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service