Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M6779

Sigma-Aldrich

Small Cardioactive Peptide B

≥97% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C52H80N14O11S2
CAS Number:
Molecular Weight:
1141.41
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥97% (HPLC)

storage temp.

−20°C

SMILES string

CSCCC(N)C(=O)NC(CC(N)=O)C(=O)NC(Cc1ccc(O)cc1)C(=O)NC(CC(C)C)C(=O)NC(C)C(=O)NC(Cc2ccccc2)C(=O)N3CCCC3C(=O)NC(CCCNC(N)=N)C(=O)NC(CCSC)C(N)=O

Amino Acid Sequence

Met-Asn-Tyr-Leu-Ala-Phe-Pro-Arg-Met-NH2

Biochem/physiol Actions

Cardioactive peptide originally isolated from Aplysia.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S L Hooper et al.
Brain research, 657(1-2), 337-341 (1994-09-19)
Application of small cardioactive peptide (SCP) or stimulation of motorneuron B15 increases the level of activated cAMP-dependent protein kinase (cAPK) in the ARC muscle. SCP application also appears to induce a translocation of cAPK between different subcellular compartments of the
S Sugita et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 17(19), 7237-7244 (1997-09-20)
The synaptic connections between the sensory neurons of Aplysia and their follower neurons have been used as a model system for examining the cellular mechanisms contributing to neuronal and synaptic plasticity. Recent studies suggest that at least two protein kinases
Beatriz P Settembrini et al.
Cell and tissue research, 321(2), 299-310 (2005-06-11)
The distribution of FMRFamide (FMRFa)-like immunoreactivity (LI) was studied in the brain and subesophageal ganglion of Triatoma infestans, the insect vector of Chagas' disease. The neuropeptide displayed a widespread distribution with immunostained somata in the optic lobe, in the anterior
L I Ismailova et al.
Biofizika, 42(4), 800-805 (1997-07-01)
Theoretical conformational analysis was carried out for the cardioactive nonapeptide Met1-Met9-NH2. Calculations yielded the values of all dihedral angles of the back bone and side chains of the nonapeptide various forms as well as intra- and interresidue interaction energies.
K Langworthy et al.
Cell and tissue research, 288(1), 191-206 (1997-04-01)
The distributions of serotonin- (5HT-), substance P- (SP-), small cardioactive peptideb- (SCPb-), and histamine- (HA-) like immunoreactivities were examined in the adult lobster supraesophageal ganglion. Vibratome sections were labeled using avidin-biotin-peroxidase immunocytochemical methods. The localization patterns for each substance were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service