Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M6692

Sigma-Aldrich

Monoclonal Anti-Minor Vault Protein 193 antibody produced in mouse

250 μg/mL, clone p193-4, tissue culture supernatant

Synonym(s):

Anti-VPARP, Anti-p193

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

tissue culture supernatant

antibody product type

primary antibodies

clone

p193-4, monoclonal

species reactivity

human

concentration

250 μg/mL

technique(s)

immunocytochemistry: 1:10-1:20 using 4% paraformaldehyde-fixed cytospin preparation
immunohistochemistry (frozen sections): 1:10-1:20
western blot (chemiluminescent): 1:10-1:20

isotype

IgG1κ

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... PARP4(143)

Looking for similar products? Visit Product Comparison Guide

General description

Minor vault protein 193 is also known as poly (ADP-ribose) polymerase family member 4 (PARP4). Along with two other proteins, it is part of vaults (ribonucleoprotein particles present in the cytoplasm). The molecular weight of this protein is 193kDa.

Specificity

Reacts with an internal epitope of (aa 491-494, HPGE) of p193.

Immunogen

E. coli lysate transformed with the pET28a(+) expression vector containing amino acids 408-611 of p193 cDNA.

Biochem/physiol Actions

Minor vault protein 193 is highly expressed in tumors.

Physical form

Supplied in serum-free culture medium containing 0.7% bovine serum albumin and 0.1% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The 193-kD vault protein, VPARP, is a novel poly(ADP-ribose) polymerase.
Kickhoefer VA
The Journal of Cell Biology, 146(5), 917-928 (1999)
Formate assay in body fluids: application in methanol poisoning.
A B Makar et al.
Biochemical medicine, 13(2), 117-126 (1975-06-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service