Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

M6628

Sigma-Aldrich

O-Methylserotonin hydrochloride

crystalline

Synonym(s):

3-(2-Aminoethyl)-5-methoxyindole hydrochloride, 5-Methoxytryptamine hydrochloride, Mexamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H14N2O · HCl
CAS Number:
Molecular Weight:
226.70
Beilstein:
3717598
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

form

crystalline

mp

245-250 °C (dec.) (lit.)

storage temp.

2-8°C

SMILES string

Cl.COc1ccc2[nH]cc(CCN)c2c1

InChI

1S/C11H14N2O.ClH/c1-14-9-2-3-11-10(6-9)8(4-5-12)7-13-11;/h2-3,6-7,13H,4-5,12H2,1H3;1H

InChI key

TXVAYRSEKRMEIF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Nonselective serotonin receptor agonist that lacks affinity for the 5-HT3 receptor.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phedias Diamandis et al.
Nature chemical biology, 3(5), 268-273 (2007-04-10)
The identification of self-renewing and multipotent neural stem cells (NSCs) in the mammalian brain holds promise for the treatment of neurological diseases and has yielded new insight into brain cancer. However, the complete repertoire of signaling pathways that governs the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service